Tisk…

Anti-PDGF-BB Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5489-100EA 6910 CZK
BIOV5489-100
Anti-PDGF-BB Rabbit Polyclonal Antibody
Protilátky
Mouse Platelet Derived Growth Factor (PDGF) is a potent mitogen for a wide range of cell types including fibroblasts, smooth muscle and connective tissue. PDGF-BB is a 24.4.0 kDa protein containing 109 amino acid residues. The reduced form of PDGF-BB shows ~14-18 kDa on SDS-PAGE.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: PDGF-BB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR