Tisk…

Anti-FABP1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5501-200EA 6910 CZK
BIOV5501-200
Anti-FABP1 Rabbit Polyclonal Antibody
Protilátky
Human Fatty Acid Binding Protein-s exhibit high affinity for small lipophilic ligands. Studies sμggest that FABPs are involved in the uptake and metabolism of fatty acids, maintenance of cellular membrane fatty acids levels, intracellular trafficking, modulation of specific enzymes of lipid metabolic pathways, as well as in the modulation of cell growth and differentiation.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml;

Type: Primary
Antigen: FABP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR