Tisk…

Anti-SLC Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5534-100EA 7470 CZK
BIOV5534-100
Anti-SLC Rabbit Polyclonal Antibody
Protilátky
SLC (secondary lymphoid-tissue chemokine, also called 6Ckine, TCA4, Exodus-2) is a 111-amino acid (human; 110 a.a. for mouse) CC chemokine with an N-terminal sequence of 23 residues as a signal sequence. It was originally cloned in 1997. Recombinant SLC attracts T-, B- cells, mature dendritic cells, but did not induce any cell migration in neutrophils or monocytes. SLC is a functional ligand for CCR7.

Recommended Dilutions: Western Blot: 0.5-1 µg/ml

Type: Primary
Antigen: SLC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR