Tisk…

Anti-C3aR Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5543-100EA 7470 CZK
BIOV5543-100
Anti-C3aR Rabbit Polyclonal Antibody
Protilátky
C3a is an anaphylatoxin generated throμgh activation of the complement C3. C3a is a potent chemoattractant for phagocytes, and it stimulates chemotaxis and other leukocyte functions. C3a binds to eosinophils, basophils, neutrophils, and differentiated U937 cells, that all express receptors for C3a on their cell surface. The C3a receptor has been cloned and shown to be a G protein-coupled receptor with seven putative transmembrane domains. This receptor is characterized for its large extracellular loop between the fourth and fifth transmembrane domains. The loop can be readily detected using the rabbit anti-C3a receptor antibody.

Recommended Dilutions:

Type: Primary
Antigen: C3aR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR