Tisk…

Anti-IL-19 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5546-100EA 7780 CZK
BIOV5546-100
Anti-IL-19 Rabbit Polyclonal Antibody
IL-19 is a single, non-glycosylated polypeptide with 152 amino acids. It plays an important role in regulating development and functioning of the skin .

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: IL-19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR