Tisk…

Anti-Resistin Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5561-100EA 6910 CZK
BIOV5561-100
Anti-Resistin Rabbit Polyclonal Antibody
Protilátky
Resistin is a new member to the family of adipocyte-secreted proteins. The biological functions of Resistin as well as its molecular target are largely unknown. Whether it promotes or antagonizes insulin action is still unclear. Recombinant rat Resistin is an 11.9 kDa protein consisting of 94 amino acid residues.

Recommended Dilutions: Western Blot: 1-4 µg/ml;

Type: Primary
Antigen: Resistin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR