Tisk…

Anti-TACE Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3222R-100EA 8430 CZK
BIOV3222R-100
Anti-TACE Rabbit Polyclonal Antibody
TACE (TNF-alpha converting enzyme, ADAM17) is a membrane-bound Metalloprotease from the ADAM (a disintegrin and Metalloprotease) family of proteins. It cleaves 26kDa type II membrane-bound precursor of TNF-a into 17 kDa mature form which is secreted. TACE is also known to process other membrane proteins including TNF receptor, TGF-alpha and amyloid protein precursor. Recombinant active untagged TACE amino acid sequence is from Arg 215 to Asn 651.

Recommended Dilutions: Western Blot: 1:200

Type: Primary
Antigen: TACE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR