Tisk…

Anti-NAIP Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3228-100EA 8430 CZK
BIOV3228-100
Anti-NAIP Rabbit Polyclonal Antibody
NAIP (neuronal apoptosis inhibitor protein) is a member of the IAPs (inhibitor of apoptosis proteins) that function in cell death pathways to inhibit apoptosis. NAIP was discovered based on its association with a neurodegenerative disorder. Mutations and deletions in the NAIP gene locus is a contributing factor in spinal muscular atrophy. Coexpression of NAIP and hippocalcin protects neurons against calcium-induced cell death. NAIP is strongly expressed in anterior horn and motor cortex neurons of the normal brain. It is also found in human fetal neurons and in adult choroid plexus cells.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: NAIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR