Tisk…

Anti-JMJD1A Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3273-100EA 7780 CZK
BIOV3273-100
Anti-JMJD1A Rabbit Polyclonal Antibody
JMJD1A (Jumonji domain containing 1A), belongs to the JHDM2 histone demethylase family. Histone demethylase specifically demethylates 'Lys-9' of histone H3, thus play a central role in histone code. JMJD1A is involved in spermatogenesis by regulating expression of target genes such as PRM1 and TMP1 that are required for proper packaging and condensation of sperm chromatin.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: JMJD1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR