Tisk…

Anti-RGS3 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3280-100EA 7470 CZK
BIOV3280-100
Anti-RGS3 Rabbit Polyclonal Antibody
Protilátky
RGS3 (regulator of G protein signaling 3) is a 519-residue protein that is widely expressed in many tissues and cells. The protein contains an RGS domain near the carboxy terminus that serves as a GTPase-activating protein (GAP;RGS3 negatively regulates the activation of Giα and Gαq. A truncated RGS3 (RGS3T), with its N-terminal 313 residues removed, has been shown to negatively regulate acenylyl cyclase and PLC. RGS3T may be localized in the nucleus and RGS3 is found in cytoplasm. Translocation to membrane is important for the RGS proteins to interact with heterotrimeric G proteins.

Recommended Dilutions: Western Blot: 1 µg/ml;

Type: Primary
Antigen: RGS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR