Tisk…

Anti-TSP1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3283-100EA 8740 CZK
BIOV3283-100
Anti-TSP1 Rabbit Polyclonal Antibody
Thrombospondin-1 is believed to play a role in cell migration and proliferation, during embryogenesis and wound repair. It has also been used as a potential regulator of tumor growth and metastasis. TSP expression is highly regulated by different hormones and cytokines and is developmentally controlled. TSP stimulates the growth of vascular smooth muscle cells and human foreskin fibroblasts. A combination of IFN-gamma and TNF-alpha inhibits TSP production in these cells.

Recommended Dilutions: Western Blot: 1:200

Type: Primary
Antigen: TSP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR