Tisk…

Anti-PAI-1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5579-100EA 8430 CZK
BIOV5579-100
Anti-PAI-1 Rabbit Polyclonal Antibody
PAI-1 (plasminogen activator inhibitor-1) is originally cloned from human endothelial cell and rat hepatoma cell cDNA libraries. As a member of the serpin family of serine protease inhibitors, PAI-1 inhibits both tissue-type palsminogen activator (t-PA) and urokinase-type plasminogen activator (u-PA;High PAI-1 levels are associated with an increased risk of thromboembolic disease while PAI-1 deficiency may represent an inherited autosomal recessive bleeding disorder.

Recommended Dilutions: Western Blot: 1-2 µg/ml

Type: Primary
Antigen: PAI-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR