Tisk…

Anti-BMP-10 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5581-100EA 7780 CZK
BIOV5581-100
Anti-BMP-10 Rabbit Polyclonal Antibody
BMPs (bone morphogenetic proteins) belong to the TGF-β superfamily of structurally related signaling proteins. As implied by their name, BMPs promote and regulate bone development, growth, remodeling and repair, in both prenatal development and postnatal growth of eye, heart, kidney, skin, and other tissues. BMP-10 is essential for the proper development of the heart, but is not expressed until after cardiac patterning or looping is completed.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: BMP-10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR