Tisk…

Anti-IFN alpha 2b Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5595-100EA 8430 CZK
BIOV5595-100
Anti-IFN alpha 2b Rabbit Polyclonal Antibody
IFN-alpha 2b is a lymphoid factor that possesses potent anti-viral activity. Human IFN-a is a 19.3 kDa protein containing 165 amino acid residues.

Recommended Dilutions: Western Blot: 1-10 µg/ml

Type: Primary
Antigen: IFN alpha 2b
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR