Tisk…

Anti-IFN alpha Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5595R-100EA 6910 CZK
BIOV5595R-100
Anti-IFN alpha Rabbit Polyclonal Antibody
Protilátky
IFN-α is a lymphoid factor that possesses potent anti-viral activity. Human IFN-α is a 19.3 kDa protein containing 165 amino acid residues.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: IFN alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR