Tisk…

Anti-ADH Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5605-100EA 6910 CZK
BIOV5605-100
Anti-ADH Rabbit Polyclonal Antibody
Protilátky
ADH (alcohol dehydrogenase) is family of proteins that metabolizes a variety of substances such as ethanol, retinal, other aliphatic alcohol, hydroxysteroids, and lipid peroxidation products. With the coenzyme NAD, ADH catalyzes the reversible conversion of organic alcohol to ketones or aldehydes. ADH plays a major role in ethanol metabolism.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: ADH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: All Species
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR