Tisk…

Anti-LIGHT Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5607-100EA 8430 CZK
BIOV5607-100
Anti-LIGHT Rabbit Polyclonal Antibody
LIGHT is a type II membrane protein belonging to the TNF family of membrane-anchored ligands. It is expressed in splenocytes, activated PBL, CD8+ tumor infiltrating lymphocytes, granulocytes and monocytes. LIGHT has the ability to activate NFkB, stimulate proliferation of lymphocytes, and induce apoptosis in certain human tumor cells. LIGHT binds to three receptors; the herpes virus entry mediator type A receptor (HEVM), LTbR and a decoy receptor (DcR3;

Recommended Dilutions: Western Blot: 0.5-1 µg/ml

Type: Primary
Antigen: LIGHT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR