Tisk…

Anti-VEGF-C Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5634-100EA 7780 CZK
BIOV5634-100
Anti-VEGF-C Rabbit Polyclonal Antibody
Vascular Endothelial Growth Factor-C (VEGF-C) is a 125 amino acid protein that plays an important role in angiogenesis and many other biological processes. Human VEGF-C exhibits about 85% homology with murine VEGF-C.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml;

Type: Primary
Antigen: VEGF-C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR