Tisk…

Anti-TCR alpha Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5648-100EA 7900 CZK
BIOV5648-100
Anti-TCR alpha Rabbit Polyclonal Antibody
Protilátky
TCR (T cell antigen receptor) recognizes foreign antigens and translates such recognition events into intracellular signals that elicit a change in the cell from a dormant to an activated state. TCR is a heterodimer composed of either α and β or γ and δ chains. The vast majority of circulating T cells (95%) express the α/β heterodimer while roμghly 2-5% express the γ/δ heterodimer. CD3 chains and the CD4 or CD8 coreceptors are also required for efficient signal transduction throμgh the TCR.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: TCR alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR