Tisk…

Anti-PTHrP Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5652-100EA 7780 CZK
BIOV5652-100
Anti-PTHrP Rabbit Polyclonal Antibody
Parathyroid Hormone-like related Protein (PTHrP) is a member of the Parathyroid hormone family that is expressed in the mammary gland. It is known as a potent inhibitor of chondrocyte maturation. PTHrP is expressed by periarticular perichondrial cells and exerts its biological activity on PP-R expressing cells. During endochondral bone development PTHrP plays a major role in suppressing the onset of hypertrophic differentiation. It is also responsible for most cases of humoral hypercalcemia of malignancy. The antibody recognizes ~9.8 kDa of human PTHrP protein.

Recommended Dilutions: Western Blot: 0.5-4 ug/ml

Type: Primary
Antigen: PTHrP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR