Tisk…

Anti-Gremlin-2 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5657-100EA 6910 CZK
BIOV5657-100
Anti-Gremlin-2 Rabbit Polyclonal Antibody
Protilátky
Gremlin (also known as Increased in High Glucose Protein 2, IHG-2, Down-regulated in Mos-transformed cells protein, Drm) functions as a bone morphogenetic protein (BMP) antagonist. It acts by binding to, and forming heterodimers with BMP-2, BMP-4, BMP-7, thus preventing them from interacting with their cell surface receptors.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Gremlin-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR