Tisk…

Anti-BMP-2 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5672-100EA 8430 CZK
BIOV5672-100
Anti-BMP-2 Rabbit Polyclonal Antibody
BMPs (bone morphogenetic proteins) belong to the TGF-beta superfamily of structurally related signaling proteins. Members of this superfamily are widely represented throμghout the animal kingdom and have been implicated in a variety of developmental processes. Proteins of the TGF-beta superfamily are disulfide-linked dimmers composed of two 12-15 kDa polypeptide chains. As implied by their name, BMPs initiate, promote and regulate bone development, growth, remodeling and repair. In addition to its osteogenic activity, BMP-2 plays an important role in cardiac morphogenesis. It is also expressed in a variety of tissues such as lung, spleen, brain, liver, prostate ovary and small intestine.

Recommended Dilutions: Western Blot: 0.5-2 µg/ml

Type: Primary
Antigen: BMP-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR