Tisk…

Anti-VENTX Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3286-100EA 6910 CZK
BIOV3286-100
Anti-VENTX Rabbit Polyclonal Antibody
Protilátky
VENTX is a member of the Vent family of homeodomain proteins. VENTX may function as a transcriptional repressor and involved in mesodermal patterning and hemapoietic stem cell maintenance.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: VENTX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Xenopus
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR