Tisk…

Anti-ASC Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3291-100EA 7780 CZK
BIOV3291-100
Anti-ASC Rabbit Polyclonal Antibody
ASC (also called TMS1) is a CARD domain containing protein. ASC induces apoptosis throμgh activation of caspase-9 and inhibiting the survival of human breast cancer cells. Overexpression of ASC/TMS1 induced DNA fragmentation. ASC/TMS1 is expressed in a variety of human and mouse tissues.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: ASC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR