Tisk…

Anti-PID/MTA2 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3294-100EA 7470 CZK
BIOV3294-100
Anti-PID/MTA2 Rabbit Polyclonal Antibody
Protilátky
PID/MTA2, also termed MTA1-L1, is a novel molecule that has been found to be involved in p53 pathway. PID/MTA2 modulates the enzymatic activity of the histone deacetylase complex and its expression reduces the levels of acetylated p53. Deacetylastion of p53 by PID/MTA2 represses p53-dependent transcriptional activation and modulates p53-mediated cell growth arrest and apoptosis. PID/MTA2 is ubiquitously expressed in human tissues.

Recommended Dilutions: Western Blot: 1 µg/ml

Type: Primary
Antigen: PID/MTA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR