Tisk…

Anti-Chk2 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3305-100EA 7470 CZK
BIOV3305-100
Anti-Chk2 Rabbit Polyclonal Antibody
Protilátky
Chk2 is the mammalian homologue of yeast Cds1/Rad53. In response to DNA damage, a checkpoint kinase ATM phosphorylates and activates Chk2, which in turn phosphorylates and activates p53. Chk2 also phosphorylates and activates BRCA1, the product of a tumor suppressor gene that is mutated in breast and ovarian cancer.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; immunofluorescence: 5-10 µg/ml

Type: Primary
Antigen: Chk2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR