Tisk…

Anti-BMF Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3334R-100EA 6910 CZK
BIOV3334R-100
Anti-BMF Rabbit Polyclonal Antibody
Protilátky
BMF (for Bcl-2-modifying factor) is a novel BH3-only protein recently identified in human and mouse. The BH3 domain in BMF is required for both binding to Bcl-2 proteins and for triggering apoptosis. In healthy cells, BMF associates with the dynein light chain 2 (DLC2) component of the myosin V motors and is sequestered by the cell's actin cytoskeleton. Disruption of the actin cytoskeleton, either by depolymerization of actin filaments or by detachment of cells from the extracellular matrix, triggers release and activation of BMF, initiating the downstream apoptosis program.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 5 µg/ml

Type: Primary
Antigen: BMF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR