Tisk…

Anti-ILP-2 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3337-100EA 6910 CZK
BIOV3337-100
Anti-ILP-2 Rabbit Polyclonal Antibody
Protilátky
ILP-2 (for IAP-like protein-2) is a novel member of in the IAP (Inhibitor of Apoptosis) protein family. ILP-2 has high homology to ILP-1, but is encoded by a distinct gene that is solely expressed in testis of tested normal human tissues. ILP-2, unlike ILP-1, has no inhibitory effect on Fas and TNF induced apoptosis, but potently inhibit apoptosis induced by overexpression of Bax or by coexpression of caspase-9 with Apaf-1. ILP-2 interacts with the processed caspase-9.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml)

Type: Primary
Antigen: ILP-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR