Tisk…

Anti-Livin Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3338-100EA 7470 CZK
BIOV3338-100
Anti-Livin Rabbit Polyclonal Antibody
Protilátky
Livin is a novel member in the IAP (inhibitor of apoptosis) protein family. Livin contains a single baculoviral IAP repeat (BIR) domain and a RING finger domain and has two isoforms termed Livin-α and Livin-β. Transfection of Livin protected cells from apoptosis induced by FADD, BAX, RIP, RIP3 and DR6. Livin directly reacts with several caspases including Caspase-3, -7, and –9, and it inhibits the activation of caspase-9 induced by Apaf-1, cytochrome c and ATP.

Recommended Dilutions: Western Blot: 2-6 µg/ml)

Type: Primary
Antigen: Livin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR