Tisk…

Anti-PUMA/bbc3 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3339-100EA 8430 CZK
BIOV3339-100
Anti-PUMA/bbc3 Rabbit Polyclonal Antibody
PUMA (p53 upregulated modulator of apoptosis) is one of the pro-apoptotic Bcl-2 family members which are also transcriptional targets of p53. PUMA gene encodes two BH3 domain-containing proteins termed PUMA-α and PUMA-β. PUMA proteins bind Bcl-2, localize to the mitochondria, and induce cytochrome c release and apoptosis in response to p53. PUMA may be a direct mediator of p53-induced apoptosis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Iimmunofluorescence: 2-4 µg/ml)

Type: Primary
Antigen: PUMA/bbc3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR