Tisk…

Anti-PCNA Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3350R-100EA 7900 CZK
BIOV3350R-100
Anti-PCNA Rabbit Polyclonal Antibody
Protilátky
PCNA (Proliferating Cell Nuclear Antigen) is a 36 kDa protein also known as cyclin. In early S phase, PCNA has a very granular distribution and is absent from the nucleoli. At late S phase, PCNA is prominent in the nucleoli. There are two forms of PCNA protein present in cells, a soluble form sensitive to organic fixation and not involved in replication, and the other form is insoluble and is associated with ongoing DNA synthesis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: PCNA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR