Tisk…

Anti-HDAC6 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3606-100EA 8910 CZK
BIOV3606-100
Anti-HDAC6 Rabbit Polyclonal Antibody
Human HDAC6 (1215 a.a. residues) possesses two separate putative catalytic domains, both of which are fully functional and contribute independently to the overall activity of HDAC6. A very potent NES is present at the amino-terminus of HDAC6, which was found to play an important role in regulating the shuttling of HDAC6 protein between cytoplasm and nucleus. The shuttling process may be a critical regulatory mechanism of HDAC6 function. HDAC6 may participate in coordinating expression of a group of genes involved in the remoldeling of chromatin during cell differentiation.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 20 µg/ml

Type: Primary
Antigen: HDAC6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR