Tisk…

Anti-HDAC7 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3607-100EA 8430 CZK
BIOV3607-100
Anti-HDAC7 Rabbit Polyclonal Antibody
Human HDAC7 is composed of 912 amino acid residues. Althoμgh HDAC7 is localized mostly to the cell nucleus, it is also found in the cytoplasm, sμggesting nucleo-cytoplasmic shuttling. The histone deacetylase activity of HDAC7 maps to a carboxy-terminal domain and is dependent on interaction with class I HDACs in the nucleus. HDAC7 catalyzes removal of acetyl-groups from acetyl-lysines of histones and promotes compaction of chromatin in these regions, leading to the inhibition of gene transcription.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 20-40 µg/ml; Immunohistochemistry: 20-40 µg/ml

Type: Primary
Antigen: HDAC7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR