Tisk…

Anti-HDAC8 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3608-100EA 7470 CZK
BIOV3608-100
Anti-HDAC8 Rabbit Polyclonal Antibody
Protilátky
Mammalian HDAC-8, isolated from human kidney, is a member of the histone deacetylase family. HDAC-8 shares homology with other HDACs in the family, but has different tissue distribution. HDAC-8 is localized to the nucleus and plays a role in the tissue development as well as cancer etiology.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 10-20 µg/ml; and Immunohistochemistry: 10-20 µg/ml

Type: Primary
Antigen: HDAC8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR