Tisk…

Anti-HDAC10 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3610-100EA 7470 CZK
BIOV3610-100
Anti-HDAC10 Rabbit Polyclonal Antibody
Protilátky
HDAC family are divided into two classes, I and II. Class I of the HDAC family comprises four members, HDAC-1, 2, 3, and 8. Class II of the HDAC family comprises HDAC-4, 5, 6, and 7, the molecular weights of which are all about two-fold larger than those of the class I members. Human HDAC-1, 2 and 3 were expressed in various tissues, but the others (HDAC-4, 5, 6, and 7) showed tissue-specific expression patterns. These results sμggest that each member of the HDAC family exhibits a different, individual substrate specificity and function in vivo.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: HDAC10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR