Tisk…

Anti-HSD17B4/MPF-2 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3617-100EA 6910 CZK
BIOV3617-100
Anti-HSD17B4/MPF-2 Rabbit Polyclonal Antibody
Protilátky
HSD17B4 (17-beta-hydroxysteroid dehydrogenase 4) is a bifunctional enzyme that is involved in the peroxisomal beta-oxidation pathway for fatty acids. HSD17B4 catalyze the formation of 3-ketoacyl-CoA intermediates from both straight-chain and 2-methyl-branched-chain fatty acids.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: HSD17B4/MPF-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR