Tisk…

Anti-Uroplakin III Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3620-100EA 7760 CZK
BIOV3620-100
Anti-Uroplakin III Rabbit Polyclonal Antibody
Protilátky
Uroplakin IIIa is a component of the asymmetric unit membrane (AUM); a highly specialized biomembrane elaborated by terminally differentiated urothelial cells. It may play an important role in AUM-cytoskeleton interaction in terminally differentiated urothelial cells. It also contributes to the formation of urothelial glycocalyx which may play an important role in preventing bacterial adherence. Defects in UPK3A are a cause of renal adysplasia (RADYS) also known as renal agenesis or renal aplasia. Renal agenesis refers to the absence of one (unilateral) or both (bilateral) kidneys at birth. Bilateral renal agenesis belongs to a group of perinatally lethal renal diseases, including severe bilateral renal dysplasia, unilateral renal agenesis with contralateral dysplasia and severe obstructive uropathy.

Recommended Dilutions: Western Blot: 1:200

Type: Primary
Antigen: Uroplakin III
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR