Tisk…

Anti-Histone H4 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3624-100EA 7470 CZK
BIOV3624-100
Anti-Histone H4 Rabbit Polyclonal Antibody
Protilátky
The nucleosome is made up of four core histone proteins (H2A, H2B, H3 and H4) and is the primary building block of chromatin. The N-terminal tail of core histones undergoes different posttranscriptional modification including acetylation, phosphorylation and methylation. These modifications occur in response to cell signal stimuli and have a direct effect on gene expression.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; in Immunoprecipitation: 20-40 µg/ml; Immunohistochemistry: 20-40 µg/ml

Type: Primary
Antigen: Histone H4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR