Tisk…

Anti-ROBO4 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3625-100EA 8430 CZK
BIOV3625-100
Anti-ROBO4 Rabbit Polyclonal Antibody
ROBO4 contains two fibronectin type-III domains and two Ig-like C2-type domains. ROBO4 is predominantly expressed in vascular endothelium and is considered important for vascular development. ROBO4 is a receptor for Slit proteins and may mediate the inhibition of primary endothelial cell migration by Slit proteins.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: ROBO4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR