Tisk…

Anti-Vimentin Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3634-100EA 8430 CZK
BIOV3634-100
Anti-Vimentin Rabbit Polyclonal Antibody
Vimentin, a intermediate filament protein, is a general marker of cells originating in the mesenchyme. Vimentin is frequently co-expressed with other members of the intermediate filament family such as the cytokeratins, in neoplasm.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 10-20 µg/ml; Immunohistochemistry: 10-20 µg/ml

Type: Primary
Antigen: Vimentin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine, Chicken
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR