Tisk…

Anti-Cox-4 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3638-100EA 8430 CZK
BIOV3638-100
Anti-Cox-4 Rabbit Polyclonal Antibody
Cytochrome c oxidase (COX) is a 13-subunit complex spanning the inner mitochondrial membrane and responsible for the terminal reduction of dioxygen to water in the electron transport chain. The three core catalytic units COX-1, -2, -3 are trans-membrane proteins encoded by the mitochondrial genome, whereas the remaining 10 subunits are nuclear encoded and expressed in a tissue-specific manner. The expression of nuclear and mitochondrial subunits of the mitochondrial respiratory chain is thoμght to be highly coordinated. Cox-4 is believed to regulate COX activity according to the extramitochondrial ATP/ADP ratio.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 10-20 µg/ml

Type: Primary
Antigen: Cox-4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR