Tisk…

Anti-BMP-3 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5673R-100EA 7470 CZK
BIOV5673R-100
Anti-BMP-3 Rabbit Polyclonal Antibody
Protilátky
BMPs (bone morphogenetic proteins) belong to the TGF-beta superfamily of structurally related signaling proteins. Members of this superfamily are widely represented throμghout the animal kingdom and have been implicated in a variety of developmental processes. Proteins of the TGF-beta superfamily are disulfide-linked dimmers composed of two 12-15 kDa polypeptide chains. As implied by their name, BMPs initiate, promote and regulate bone development, growth, remodeling and repair. In vivo studies have shown that BMP-3 can independently induce cartilage formation.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: BMP-3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR