Tisk…

Anti-BMP-4 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5674R-100EA 6910 CZK
BIOV5674R-100
Anti-BMP-4 Rabbit Polyclonal Antibody
Protilátky
BMPs (bone morphogenetic proteins) belong to the TGF-beta superfamily of structurally related signaling proteins. Members of this superfamily are widely represented throμghout the animal kingdom and have been implicated in a variety of developmental processes. Proteins of the TGF-beta superfamily are disulfide-linked dimmers composed of two 12-15 kDa polypeptide chains. As implied by their name, BMPs initiate, promote and regulate bone development, growth, remodeling and repair. Smad1 translocation to the nucleus is observed after the addition of BMP-4 (also designated BMP-2B), sμggesting that BMP-4 may play a role in activation of the Smad pathway.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: BMP-4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Chicken, Rabbit
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR