Tisk…

Anti-Galectin-3 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5684-100EA 6910 CZK
BIOV5684-100
Anti-Galectin-3 Rabbit Polyclonal Antibody
Protilátky
Lectins are carbohydrate binding proteins that interact with glycoprotein and glycolipids on the surface of animal cells. The Galectins are lectins that recognize and interact with beta-galactoside moieties. Galectin-3 regulates a number of biological processes, including embryogenesis, inflammatory responses, cell progression and metastasis. Galectin-3 can function intracellularly in controlling cell cycle and preventing T-cell apoptosis, and also extracellularly, in activating various cells, including monocytes/macrophages, mast cells, neutrophils, and lymphocytes. Human Galectin-3 is a globular 26.0 kDa protein containing 250 amino acid residues.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Galectin-3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR