Tisk…

Anti-ApoA1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5693-100EA 7780 CZK
BIOV5693-100
Anti-ApoA1 Rabbit Polyclonal Antibody
Apolipoprotein A-I (ApoA1) is the primary protein constituent of HDL, defining its size and shape, solubilizing its lipid components, removing cholesterol from peripheral cells, activating the LCAT enzyme, and delivering the resulting cholesterol esters to the liver.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: ApoA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR