Tisk…

Anti-IGF-BP1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5717-100EA 6910 CZK
BIOV5717-100
Anti-IGF-BP1 Rabbit Polyclonal Antibody
Protilátky
Human IGF-BP1 is a cysteine-rich secreted protein expressed in liver, decidual, and kidneys and is the most abundant IGF-BP in amniotic fluid. Levels of IGF-BP1 in serum are lowest after food. IGF-BP1 binds both IGF-I and IGF-II with equal affinity. Phosphorylated IGF-BP1 hinders IGF actions, whereas nonphosphorylated IGF-BP1 is stimulatory.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: IGF-BP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR