Tisk…

Anti-IL-12p40 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5732-100EA 6910 CZK
BIOV5732-100
Anti-IL-12p40 Rabbit Polyclonal Antibody
Protilátky
Interleukin-12 is a disulfide-linked heterodimeric protein (p70), composed of two subunits, p35 and p40, which are encoded by two different genes. Recombinant Human IL-12 p40 produced in E.Coli is a single, non-glycosylated polypeptide chain containing 306 amino acids fragment (23-328;

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: IL-12p40
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR