Tisk…

Anti-PlGF Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5743-100EA 7470 CZK
BIOV5743-100
Anti-PlGF Rabbit Polyclonal Antibody
Protilátky
PLGF (also called Placenta Growth Factor, PIGF, PGF) is an angiogenic factor that belongs to the cysteine-knot superfamily of growth factors. PLGF is expressed in placental tissues, colon and mammary carcinomas. It signals throμgh the VEGFR-1/FLT1 receptor and stimulates endothelial cell proliferation and migration. Recombinant mouse PLGF is a disulfide-linked homodimer containing two 142 amino acid polypeptide chains.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: PlGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR