Tisk…

Anti-TFF-2/Spasmolysin Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5745-100EA 6910 CZK
BIOV5745-100
Anti-TFF-2/Spasmolysin Rabbit Polyclonal Antibody
Protilátky
The Trefoil Factor peptides (TFF1, TFF2 and TFF3) are expressed in the gastrointestinal tract, and appear to play an important role in intestinal mucosal defense and repair. TFF2 has been shown to inhibit gastrointestinal motility and gastric acid secretion. Recent data sμggests a potential role for TFF2 in acute and chronic asthma. Human TFF2 is a 12.0 kDa polypeptide of 106 amino acid residues.

Recommended Dilutions: Western Blot: 0.5-2 µg/ml

Type: Primary
Antigen: TFF-2/Spasmolysin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR