Tisk…

Anti-Urokinase Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5793-100EA 7470 CZK
BIOV5793-100
Anti-Urokinase Rabbit Polyclonal Antibody
Protilátky
Western Blot analysis (1-4 µg/ml;However, the optimal concentrations should be determined individually.

Recommended Dilutions: N/A

Type: Primary
Antigen: Urokinase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR